Order Entry
Sweden
ContactUsLinkComponent
Anti-MD-2 Rabbit Polyclonal Antibody
Anti-MD-2 Rabbit Polyclonal Antibody
Catalog # ENZOADICSA506E
Supplier:  ENZO LIFE SCIENCES
Anti-MD-2 Rabbit Polyclonal Antibody
Catalog # ENZOADICSA506E
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Lymphocyte antigen 96
  • Antigensymbol:
    LY96
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Liquid
  • Antigen synonymer:
    ESOP-1|MD2|MD-2|ly-96|lymphocyte antigen 96
  • Storage buffer:
    PBS containing 0,02% sodium azide
  • Förvaringstemperatur:
    4 °C
  • Shipping temperature:
    blue ice not frozen
  • Immunogen:
    Synthetic peptide corresponding to the sequence near the middle of human MD-2
  • Rening:
    Purified by ion exchange chromatography
  • Storlek:
    100 μg
  • Förp.:
    100 µG

Specifications

About this item

MD-2 is a member of the Toll/interleukin-1 receptor (TIR) family, a group of proteins that include the Toll-like receptors (TLRs). TLRs recognize different pathogen-associated molecular patterns, thereby linking the innate and adaptive immune responses.

  • Dilution range: 2 µg/ml for Immunohistochemistry, 0,5 to 2,0 µg/ml for Western blot

TLR4, the major signaling receptor for lipopolysaccharide (LPS), requires the binding of MD-2 to its extracellular region for maximal response to LPS.

Type: Primary
Antigen: Lymphocyte antigen 96
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat