Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Supplier:   BOCHEM
Forceps, precision
Supplier:  BOCHEM
Certificates
Forceps, precision
Forceps, precision
3 Options Available Below
Product Details & Documents
These non serrated forceps are made of durable titanium.
  • High corrosion and thermal resistance
  • Lightweight, high strength
Titanium is resistant to diluted sulphuric acid, hydrochloric acid, solutions containing chloride, cold nitric acid as well as most organic acids and alkaline solutions, such as sodium hydroxide.
Product Family Options
Product Information
  • Beskrivning
    Typ
    Längd
    Availability
    Price
  • Precision forceps, extra sharp, straight (tower tip)
    Straight
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 765,00 kr
    765,00 kr
    1 * 1 Items
       765,00 kr
  • Precision forceps, extra sharp, curved
    Curved
    105 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 765,00 kr
    765,00 kr
    1 * 1 Items
       765,00 kr
  • Precision forceps, straight, smooth tips, extra sharp
    Rak
    130 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 765,00 kr
    765,00 kr
    1 * 1 Items
       765,00 kr