Resistor kit for conductivity meters, Orion™
ORIO1010001
- Beskrivning:Conductivity calibration resistor kit with 8-pin MiniDIN connection, for use with Orion Star and Orion Star A Series conductivity meters
- Seq:01
- Förp.:1
This resistor calibration kit can be used to verify the performance of Thermo Scientific™ Orion™ Versa Star conductivity meters, Thermo Scientific™ Orion™ Star A series conductivity meters and legacy Orion™ Star series conductivity meters.
- Conductivity calibration resistor kit with 8 pin MiniDIN connection
: Kit includes six resistors that simulate nominal conductance values between 1 µS and 100 mS.