Anti-CPN10 Rabbit Polyclonal Antibody
Leverantör: ENZO LIFE SCIENCES
Certifikat
Om denna produkt
Chaperonin 10 (Cpn10 or Hsp10), is the ~10 kDa mammalian equivalent to GroES of E. coli. Chaperonins play fundamental roles in the folding, assembly, and translocation of other proteins.
- Dilution range: Western blot (1:2,000, ECL)
- Detects a band of ~10kDa by western blot
Type: Primary
Antigen: CPN10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Guinea pig, Human, Mouse, Porcine, Rabbit, Rat, Sheep, Xenopus
Specifikationer
- Catalog No:
- ENZOADISPA110F
- ENZOADISPA110D
- Antigensymbol:
- HSPE1
- HSPE1
- Antikroppsnamn:
- Heat shock 10kDa protein 1
- Heat shock 10kDa protein 1
- Antigen synonymer:
- Chaperonin 10
- Chaperonin 10
- Antikroppstyp:
- Primär
- Primär
- Klonalitet:
- Polyclonal
- Polyclonal
- Konjugation:
- Unconjugated
- Unconjugated
- Reaktivitet:
- Pig, Rabbit, Xenopus, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Pig, Rabbit, Xenopus, Sheep, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Värd:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunKemi:
- Yes
- Yes
- Epitop:
- C-Terminal
- C-Terminal
- Immunogen:
- Synthetic peptide corresponding to the sequence near the C-terminus of human Cpn10
- Synthetic peptide corresponding to the sequence near the C-terminus of human Cpn10
- Form:
- Liquid
- Liquid
- Rening:
- Purified by Protein A affinity chromatography
- Purified by Protein A affinity chromatography
- Storage buffer:
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
- PBS, pH 7,2, containing 50% glycerol and 0,09% sodium azide
- Förvaringstemperatur:
- −20 °C
- −20 °C
- Shipping temperature:
- Shipped on blue ice
- Shipped on blue ice