Wood and plastic moisture tester, Skipper Plus
Katalog # SIMELI9012
Leverantör: Industrial Physics
Specifikationer
- Modell:Skipper Plus
- Beskrivning:Wood and plastic moisture tester
- Förp.:1
Specifikationer
Om denna produkt
Skipper Plus is a non-destructive moisture detector designed for locating damp in wood or laminated plastics such as polyester.
- Short rubber electrodes, which do not damage the surface
- Three moisture ranges for moisture in both fresh and salt water
- Developed for osmosis-control and measuring moisture before painting yachts
W×D×H: 150×80×30 mm