Single gas detector
Catalog # SIMEDM6025
Supplier: Industrial Physics
Specifications
- Parameter:O₂
- Beskrivning:Single gas detector
- Förp.:1
Specifications
About this item
This gas clip detection equipment is designed for use in the coal mining industry.
- Adjustable alarm set points
- Real-time gas reading capability
- Two-way infrared communication