ContactUsLinkComponent
Hand cleansing cream, Solopol® Classic PURE
Hand cleansing cream, Solopol® Classic PURE
  115-2345
 :  Deb-STOKO
undefined
Hand cleansing cream, Solopol® Classic PURE
  115-2345
 :  Deb-STOKO
  • Beskrivning:
    Hand cleansing cream, Solopol® Classic PURE, 2000 ml cartridge
  • Seq:
    02
  • Förp.:
    4
Traditional soap-free hand cleanser paste with scrubbing agent for heavy-duty contamination from, for example, oils, greases, carbon black, graphite, metallic dust, lubricants. For use in industrial environments.
  • Astopon® natural scrubbing agent (refined walnut shell powder) gently removes contaminants without damaging the skin
  • Contains Eucornol® skin protectant to minimise the risk of skin irritation
  • With re-fatting emoillent and Aloe vera
  • Solvent-free, colourant-free, perfume-free
 : EU Ecolabel.
 : Soft bottle may only be used with Stoko® Vario® dispensers.
 : *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904