Order Entry
Sweden
ContactUsLinkComponent
Anti-CD40 Rabbit Polyclonal Antibody
Anti-CD40 Rabbit Polyclonal Antibody
Katalog # BSBTPB10051
Leverantör:  Boster Bio
undefined
Anti-CD40 Rabbit Polyclonal Antibody
Katalog # BSBTPB10051
Leverantör:  Boster Bio

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Cd40 Molecule, Tnf Receptor Superfamily Member 5
  • Antigensymbol:
    CD40
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Gen-ID:
    958
  • Antigen synonymer:
    p50|Bp50|TNFRSF5|CDW40
  • Storlek:
    100 μg
  • Förp.:
    100 µG

Specifikationer

Om denna produkt

Rabbit IgG polyclonal antibody for Tumor necrosis factor receptor superfamily member 5(CD40) detection. Tested with WB, ELISA in Human.

Type: Primary
Antigen: CD40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human