Order Entry
Sweden
ContactUsLinkComponent
Anti-AK1 Rabbit Polyclonal Antibody
Anti-AK1 Rabbit Polyclonal Antibody
Katalog # BSBTPB10033
Leverantör:  Boster Bio
undefined
Anti-AK1 Rabbit Polyclonal Antibody
Katalog # BSBTPB10033
Leverantör:  Boster Bio

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Adenylate Kinase 1
  • Antigensymbol:
    AK1
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Gen-ID:
    203
  • Antigen synonymer:
    HTL-S-58j
  • Storlek:
    100 μg
  • Förp.:
    100 µG

Specifikationer

Om denna produkt

Rabbit IgG polyclonal antibody for Adenylate kinase isoenzyme 1(AK1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: AK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat