Concrete inspection hammer
Katalog # SIMELD0500
Leverantör: Industrial Physics
Specifikationer
- Mätområde:10 - 110 N/mm²
- Beskrivning:Concrete inspection hammer
- Förp.:1
Specifikationer
Om denna produkt
Hammer used to test the quality of concrete, as well as other construction materials, for analysing resistance.
Certifications: EN 12504-2, ASTM, C805, BS 1881.