Culture tubes, screw thread
Catalog # SCERD61251600
Supplier: DWK Life Sciences
Specifications
- Ø:16 mm
- Höjd:125 mm
- Förp.:100
Specifications
About this item
Tubes, made up of borosilicate glass, grade 3.3, with screw threads.
- Caps must be ordered separately
- White marking spot
- Very good chemical and temperature resistance