Test kits, aluminum (Testpak), CHECKIT®
Catalog # LOVI147700
Supplier: Lovibond
CAS Number:
Specifications
- Förp.:1 KIT
- Beskrivning:Aluminium test kit
- Mätområde:0 - 0,3 mg/l Al
Specifications
About this item
CHECKIT® Comparator is a compact and handy colorimetric unit which is suitable for both mobile and static analysis work. Supplied with a generous number of different colour scales, it provides the basis for a comprehensive, easy-to use colorimetric analysis system.
- Enables the use of large path lengths
- The mirror optics makes use of the view through the entire length of the cell
- Easy operation and exact reagent dosing
- Tablet reagents have minimum guaranteed shelf life of 5/10 years
Testpak is a simple and cost-effective means of extending the use of an existing CHECKIT® comparator instrument to a new test parameter.
Delivery: Each Testpak includes the required CHECKIT® disc, tablet reagents (normally for 30 tests), two cells, stirring rod and detailed instructions for the desired method.