Test Kits, chlorine and pH, Comparator 2000+
Katalog # 705-0353
Leverantör: Lovibond
CAS-nummer:
Specifikationer
- Förp.:1 KIT
- Beskrivning:Chlorine and pH test kit
- Seq:01
- Typ:AF 116A
- Mätområde:0,1 - 1,0 mg/l (Cl2);
6,8 - 8,4 pH
Specifikationer
Om denna produkt
Comparator 2000+ is a versatile modular system for testing water.
- Stable and accurate
- Robust, compact and portable
- Prism enabling colour matching
- Diffuser plate is used to measure the turbidity
- Longer path length cells be used for lower concentration analysis
The integrated prism brings the glass standards of the test discs and the coloured sample into the same field of view.
Delivery: Test kit is supplied in a robust carrying case containing 1× chlorine comparator disc, 1× pH comparator disc, 4× comparator vials, 10 ml, 13 mm path length, tablet reagents, crushing/stirring rod, vial cleaning brush and instruction sheet.