Vane wheel for heat/air flow
Catalog # BINH1003101
Supplier: 3B Scientific
Specifications
- Beskrivning:Vane wheel
- Material:Aluminium
- Förp.:1
Specifications
About this item
Vane wheel device is used for demonstrating air and heat flows generated by a burning candle, water vapour or other heat sources. Made of Aluminium and mountable on a long needle.