ADC active digital controllers
Catalog # EDWAD39591500
Supplier: EDWARDS HIGH VACUUM METTLER
Specifications
- Noggrannhet:1×10ˉ9 mbar
- Beskrivning:Active digital controllers, ADC mkII Enhanced
- Modell:ADC mkII Enhanced
- Tillåten rumstemperatur vid drift:+0...+40 °C
- Tillåten rumstemperatur vid förvaring:−30...+70 °C
- Förp.:1
Specifications
About this item
This Active Digital Controller (ADC) is a compact single gauge controller with automatic recognizes compatible Edwards gauges, loads the appropriate look-up table and displays the pressure in commonly used vacuum units.
- Bright LED display gives clear, long distance readability
- The ADC displays units in mbar, Torr, Pascal or Volts
- Simple push-button control makes operation highly intuitive
- This gauges are mainly used in laboratory and industrial applications
The enhanced controller supports two similar gauges - it has two variable hysteresis set-points which are linked to 48 V DC. 1 A changeover relays and two 0 to 10 V DC analog outputs. To aid system integration, the enhanced controller is provided with an RS232 interface. When combined with a suitable gauge, such as the Edwards APGXH Convection Pirani or Wide Range Gauge (WRG), the ADC represents a cost effective means of monitoring and controlling process vacuum in a broad range of applications.