Indirect impact tester
Catalog # SIMESP1880
Supplier: Industrial Physics
Specifications
- Beskrivning:Impact tester incl. base plate assembly, guide tube, release collar, two punches (Ø = 12,7 and 15,9 mm)
- Seq:05
- Förp.:1
Specifications
About this item
Impact testers used to determine the impact resistivity and flexibility of materials.
- Spirit level built-in for correct positioning
- Dual scale instrument equipped with a special guidance
- Distance between each impact is always according to the standards
Certifications: ISO 6272-2 and ASTM D2794