Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Omrörare
Omrörarmagneter
Magnetic stirring bars
Magnetic stirring bars
Magnetic stirring bars
Catalog # HACHLZE136
Supplier:
Hach
Expand image
Magnetic stirring bars
Catalog # HACHLZE136
Supplier:
Hach
Specifications
Ø:
6 mm
Längd:
20 mm
Förp.:
5
Specifications
About this item
Product Details
PFTE coated magnetic stirring bars.
Product Details