Crack set for metal digestion
Katalog # HACHLCW902
Leverantör: Hach
CAS-nummer:
Specifikationer
- Förp.:50
- Beskrivning:Crack set for metal digestion- suitable for cadmium, copper, lead, iron, nickel and zinc.
Specifikationer
Om denna produkt
Crack set for digestions of complex, colloidal or undissolved metals in water and waste water.