ContactUsLinkComponent
Crack set for metal digestion
Crack set for metal digestion
Katalog # HACHLCW902
Leverantör:  Hach
CAS-nummer:  
undefined
Crack set for metal digestion
Katalog # HACHLCW902
Leverantör:  Hach
CAS-nummer:  
Specifikationer
  • Förp.:
    50
  • Beskrivning:
    Crack set for metal digestion- suitable for cadmium, copper, lead, iron, nickel and zinc.
Specifikationer
Om denna produkt
Crack set for digestions of complex, colloidal or undissolved metals in water and waste water.