Supplied air regulator, Versaflo™ V-500E
111-2225
: 3M
- Beskrivning:Compressed air regulator, Versaflo™ V-500E, with belt
- Förp.:1
3M™ Versaflo™ V-Series supplied air regulators offer protection against dust, mist, fumes, gases or vapours and combination hazards e.g. paint spray. Supplied air regulators do not offer protection in confined spaces, oxygen deficient atmospheres or for asbestos stripping. The 3M™ Versaflo™ V-500E supplied air regulator is a balanced, belt-worn, easy to adjust regulator specifically designed to ensure smooth airflow, safety and comfort.
- Low pressure warning whistle
- Integrated silencer
- Individually adjustable, consistently controlled airflow
- Removable, integrated activated charcoal filter
- Belt swivel mechanism
- Spray gun or compressed air tool port
- Supplied air connection
- Components of the Versaflo™ V-Series regulators are connected using plug and socket hose couplings (Rectus 25)
Airflow: 170 to 305 L/min
: Approved to EN 14594 Class 2A/3A or 2B/3B dependent on helmet.