Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Elektrokemimätare
Syremätare
Accessories for oxygen meter GMH 3691
Accessories for oxygen meter GMH 3691
Accessories for oxygen meter GMH 3691
Catalog # GRSG601490
Leverantör:
Greisinger
Expand image
Accessories for oxygen meter GMH 3691
Catalog # GRSG601490
Leverantör:
Greisinger
Specifikationer
Beskrivning:
Spare air oxygen sensor, GOEL 370, for universal and diving applications, for gases with increased CO2 concentrations
Seq:
10
Förp.:
1
Specifikationer
Om denna produkt
Product Details
Product Details