Safety shoes, lace-up, Cosy Grey
Catalog # 114-6683
Supplier: HONEYWELL SAFETY
Specifications
- Skostorlek:39
- Färg:Grå
- Förp.:1 PAIR
Specifications
About this item
100% metal-free safety shoe for ladies made from microfibre upper, a padded ankle collar and a slip resistant injected dual-density PU/rubber outsole.
- Poromax lining ensures high breathability
- Dual-density insole for comfort
- Metal-free Smartium™ toecap made from injected polymer (200 J)
- Non-metallic puncture resistant midsole protects 100% of the foot surface
Colour: Grey
Certifications: EN ISO 20345:2011; S3, SRC, CI, HI