ContactUsLinkComponent
Beakers, glassy carbon
Beakers, glassy carbon
Katalog # 39025.KT
Leverantör #: 39025.KT
Beakers, glassy carbon
Katalog # 39025.KT
Leverantör #: 39025.KT
Logga in för tillgänglighet
Select an Option
90 mm
Select an Option
400 ml
Select an Option
86 mm
Select an Option
1
Om denna produkt
Beakers, glassy carbon.
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
Vessels for ultra-high purity materials technology, e.g. semiconductor connections and crystal­growing, e.g. doped halogenide crystals.