Decontamination bags, activated charcoal
Katalog # 22007.
Supplier: Biotium
CAS Number:
Specifications
- Förpackningstyp:bags
- Förp.:25
- Beskrivning:Activated charcoal decontamination bags
- Seq:03
Specifications
About this item
Activated charcoal retains ethidium bromide, GelRed™ and GelGreen™ efficiently in a convenient bag for decontamination of used gel staining solutions. Each bag has a total capacity of at least 10 mg ethidium bromide.
- Remove mutagenic dyes (ie, ethidium bromide) from gel staining solutions
- Use to clean up any dyes from a solution to allow drain disposal
- No-mess, convenient bag
- Reusable bag capacity of at least 10 mg EtBr