Quartz wool
ROTH9208.2
- Beskrivning:Quartz wool
- Seq:02
- Förp.:100 g
This quartz wool is ideal for elementary analysis, as a final stopper in chromatography columns or as a filter aid.
- Chemically pure
- Fibre size 5 to 30 µm
: Packed in plastic bottles.