Biopsy cassettes, disposable lid
Katalog # KLIN210-07T
Leverantör: KLINIPATH
Specifikationer
- Färg:Grå
- Seq:31
- Förp.:1.600
Specifikationer
Om denna produkt
Biopsy cassettes with hinged lid.
- Cassettes maintain their stability, rigidness and shape during processing of tissues
- Available in different colours
- Lids are packed separately
Packaging: Hoppers contain 80 cassettes and are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes and are specially packed for Sakura® / Leica® cassette printers Lasermark cassettes are specially packed for laser marking technology.