Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Chromosulphuric acid
Chromosulphuric acid
Chromosulphuric acid
:
Thermo Fisher Scientific
Chromosulphuric acid
:
Thermo Fisher Scientific
:
65272-71-1
:
:
Cr₂K₂O₇·H₂O₄S
:
MFCD00011368
:
392.25 g/mol
:
330 °C (760 mmHg)
:
1,84 g/cm³
:
Omgivande
:
2240
:
8,I
:
GHS05
:
GHS08
Appearance:
Red-brown to dark brown Liquid
Assay:
≥92 % H2SO4
ACRO29551
ACRO295510025_TDS_EN