Order Entry
Sweden
ContactUsLinkComponent
 
Anti-MGP Antibody
  BSBTPB9954
undefined
Anti-MGP Antibody
  BSBTPB9954

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    matrix Gla protein
  • Antigensymbol:
    MGP
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Gen-ID:
    4256
  • Antigen synonymer:
    MGLAP|GIG36|NTI
  • Storlek:
    100 µg
  • Förp.:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Matrix Gla protein(MGP) detection. Tested with WB, IHC-P in Human.

Type: Primary
Antigen: MGP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human