Order Entry
Sweden
ContactUsLinkComponent
Anti-ABL2 Antibody
Anti-ABL2 Antibody
Catalog # BSBTPB9913
Supplier:  Boster Bio
undefined
Anti-ABL2 Antibody
Catalog # BSBTPB9913
Supplier:  Boster Bio

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    ABL proto-oncogene 2, non-receptor tyrosine kinase
  • Antigensymbol:
    ABL2
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Unconjugated
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Korsad adsorption:
    No
  • Gen-ID:
    27
  • Antigen synonymer:
    ARG|ABLL
  • Storlek:
    100 µg
  • Förp.:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Abelson tyrosine-protein kinase 2(ABL2) detection. Tested with WB, IHC-P in Human;Mouse.

Type: Primary
Antigen: ABL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse