Immunoassay microplates, Protein L coated plates, Pierce™
Katalog # PIER15190
Leverantör: Thermo Fisher Scientific
Specifikationer
- Förp.:5 plates per pack
- Färg:Klar
- Beskrivning:96-well plates, Protein L coated, Polystyrene
Specifikationer
Om denna produkt
Pierce™ Protein L coated plates provide an alternative to direct, passive adsorption methods for immobilising antibodies for ELISA and other plate-based assay techniques.
- Retain antibody activity, which can be lost when antibodies are immobilised by passive adsorption
- Orient antibodies for maximum antigen-binding capacity
- Immobilise antibodies for plate assays without prior purification
- Ensure minimal variation (<5% well-to-well) from consistent coating
- Plates are pre-blocked with SuperBlock™ blocking buffer to reduce non specific binding
- Bind 2 to 5 pmol of antibody per well (based on tests with rabbit IgG)
- Binding Property: Standard Capacity, Affinity; Detection Method: Colorimetric
Protein L binds to immunoglobulin kappa light chains without interfering with the antigen-binding site. Protein L binds to all classes of Ig (ie. IgG, IgM, IgA, IgE and IgD). Protein L also binds single chain variable fragments (scFv) and Fab fragments.