Streptavidin, hydrazide-activated, Pierce™
Catalog # PIER21120
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Förp.:2 mg
- Beskrivning:Streptavidin, hydrazide-activated
Specifications
About this item
Pierce™ Hydrazide-activated strepavidin conjugate includes recombinant streptavidin in a purified form activated for crosslinking to carbonyl groups in a molecule.