Protein L Plus agarose, Pierce™
Katalog # PIER20520
Leverantör: Thermo Fisher Scientific
CAS-nummer:
Specifikationer
- Förpackningstyp:Bottle
- Förp.:2 ml
- Beskrivning:Protein L Plus agarose
Specifikationer
Om denna produkt
Pierce™ Protein L Plus agarose is a high-capacity Protein L beaded agarose resin for use in a variety of antibody affinity purification methods.
- Immobilised Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
- Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
- Protein L is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
- High capacity beads with a dense load of immobilised Protein L
Binding capacity: 10 to 20 mg human IgG/ml resin