Horseradish Peroxidase, Biotinylated, Pierce™
Katalog # PIER29139
Leverantör: Thermo Fisher Scientific
CAS-nummer:
Specifikationer
- Förp.:5 mg
- Beskrivning:Horseradish peroxidase, biotinylated
Specifikationer
Om denna produkt
Biotinylated horse radish peroxidase is used as control or signal amplification in IHC to amplify the signal of biotinylated primary antibodies using the avidin-biotin complex staining method.