Electrophoresis running buffers, Pierce™
Katalog # PIER28362
Leverantör: Thermo Fisher Scientific
CAS-nummer:
Specifikationer
- Förp.:1 L
- Beskrivning:Tris-Glycine SDS buffer, 10X
- Seq:02
Specifikationer
Om denna produkt
These are space-saving stock solution that is ideal for quickly preparing running buffer used for gel electrophoresis.
- Eliminates the possibility of powder remaining in a packet
- Eliminates time spent waiting for powder to dissolve
- Storage as concentrated stock minimises bench space needed for solutions
Tris-HEPES-SDS buffer - when diluted 20-fold in water, the solution yields 0,1 M Tris, 0,1 M HEPES, 3 mM SDS, pH 8
Tris-glycine-SDS buffer - when diluted 10-fold in water, the solution yields 0,025 M Tris, 0,192 M glycine, 0,1% SDS, pH 8,5