Orders LinkContactUsLinkComponent
Freund's complete adjuvant, Imject™
Freund's complete adjuvant, Imject™
Imject™ Freund's complete adjuvant, 5×10 ml, 1 SET
Katalog # PIER77140
Leverantör:   Thermo Fisher Scientific
Freund's complete adjuvant, Imject™
Katalog # PIER77140
CAS-nummer:  
Freund's complete adjuvant, Imject™
Freund's complete adjuvant, Imject™
Imject™ Freund's complete adjuvant, 5×10 ml, 1 SET
CAS-nummer:  
Price
2 675,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 SET
Product Details & Documents
Imject™ Freund's Complete Adjuvant (FCA) is a popular non specific stimulator of the immune response for use in antibody production immunisation protocols.
  • Greatly enhances immune response to immunogen
  • Typically used for initial injections
  • Water-in-oil emulsion containing killed Mycobacterium
Leveransinformation: Supplied ready to use in 10 ml ampoules.
Specifikationer
Specifikationer
  • Förp.:
    1 SET
  • Beskrivning:
    Imject™ Freund's complete adjuvant, 5×10 ml
Specifikationer