Cryopreservation media, HyClone™ HyCryo™
Katalog # SR30001.02
Leverantör: HyClone products (Cytiva)
CAS-nummer:
Specifikationer
- Förp.:100 ml
- Beskrivning:HyClone™ HyCryo cryopreservation medium
Specifikationer
Om denna produkt
Animal origin-free HyClone™ HyCryo cryopreservation media is designed for general use.
- 0,2 μm sterile filtered
- Serum free formulation
- Increased recovery of viable cells versus standard cryopreservation formulations