VWR®, Prepared Slides: Freshwater Plants (Cryptogams)
Katalog # 720-2925
Leverantör: VWR Collection
Specifikationer
- Beskrivning:Freshwater plants (cryptogams) slides
- Förp.:1 KIT
Specifikationer
Om denna produkt
Prepared slides for freshwater plants (cryptogams) including.
- Three types of bacteria smear
- Rhizopus nigricans (bread mould) (w.b.)
- Penicillium (w.b.)
- Aspergilus (w.b.)
- Yeast (w.b.)
- Lichen (s.)
- Volvox (w.b.)
- Spirogyra conjugation (w.b.)
- Porphyra (sea lettuce) (w.b.)
- Marchantia thallus (s.)
- Marchantia antheridiophore (l.s.)
- Marchantia archegoniophore (l.s.)
- Moss, protonema (w.b.)
- Moss, antheridium (c.s.)
- Fern prothallus, young sporophyte (w.b.)
(s.) = section; (l.s.) = longitudinal section; (c.s.) = cross section; (w.b.) = whole body.