VWR®, Prepared Slides: General Biology
Catalog # 720-2924
Supplier: VWR Collection
Specifications
- Beskrivning:General biology prepared slides, set 1
- Förp.:1 KIT
Specifications
About this item
Set of prepared slides for educational purposes, to better understand general biology.
- Unicellular animal i.e. amoeba or other
- Paramecium (w.b.)
- Hydra (w.b.)
- Daphnia (w.b.)
- Lumbricus, earthworm (c.s.)
- House fly mouth (w.b.)
- Honey bee mouth (w.b.)
- Honey bee hind leg
- Three bacteria smears
- Simple flat epitelium (w.b.)
- Skeletal muscle (l.s. and c.s.) human blood smear
- Frog blood smear
- Lung section
- Artery and vein (c.s.)
- Human skin section through hair follicle
- Spirogyra conjugation (w.b.)
- Volvox (w.b.)
- Moss (w.b.)
- Mitosis, onion root tip cell (s.)
- Monocot and dicot roots (c.s.)
- Monocot and dicot shafts (c.s.)
- Tilia (linden tree), 1 year old stem (c.s.)
- Tilia (linden tree), 3 year old stem (c.s.)
- Monocot and dicot leaves (c.s.)
(c.s.) = cross section; (l.s.) longitudinal section; (w.b.) = whole body.