Detergent, alkaline, ClearKlens® Pro VH17
Catalog # 141-1376
Supplier: Diversey
Specifications
- Beskrivning:ClearKlens® Pro VH17 detergent
- Förp.:20 L
- pH-område:11 - 13 (1% solution)
Specifications
About this item
ClearKlens Pro is a low foaming, soft metal safe, hard water tolerant alkaline silicated detergent for use in CIP and spray washing applications. ClearKlens Pro has also been tested and approved for compatibility and use on polycarbonate.
- ClearKlens Pro is safe for use on polycarbonate moulds, aluminium and other soft metal surfaces
- Highly versatile and efficient detergent
- The surfaces remain free from corrosion and pitting corrosion and the service life of the equipment is extended
- Low foaming under high turbulence making it easier to rinse and improving cleaning efficiency
- Hard water tolerant, preventing scale build up and improving operational efficiency
ClearKlens Pro provides excellent fat/oil penetration and removal properties for CIP and spray washing applications, for the pharmaceutical and cosmetic industries.