pH electrode, Radiometer Analytical
Catalog # HACHE11M001
Supplier: Hach
Specifications
- Anslutning:Screw cap (no cable)
- Ø×L:12×103 mm
- För:General purposes
- Typ:Laboratory pH electrode
- Beskrivning:pH electrode, PHG301-9 (sensor only, external reference probe needed)
- Material:Glas
- Förp.:1
- Temp:0...80 °C
- pH:0 - 12
Specifications
About this item
These glass electrodes are compatible with Hach sensION+ benchtop pH meters (or any meter with a BNC connection). The PHG301 pH electrode should be used with Ag/AgCl or calomel reference electrodes.
- Made with highly shock-resistant glass, making them extremely tough
- Average lifetime of a correctly maintained Radiometer glass pH electrode is approximately two years
The PHG301 is a screw cap electrode and requires the CL114 cable for connection to a meter.
Delivery: Supplied with certificate of conformity.