Electrode filling solution, IntelliCAL™ PHC281
Katalog # HACH2965026
Leverantör: Hach
CAS-nummer:
Specifikationer
- Förp.:59 mL
- Beskrivning:Internal filling solution for use with PHC281 IntelliCAL™ Ultra pH probes, 2,44M KCl, 59 ml bottle with dispenser cap