Vatten cell culture reagent DNase-, RNase-free
Catalog # ICNA04821739
Supplier: MP Biomedicals
CAS Number:
Specifications
- Förp.:500 mL
- Seq:2725
Specifications
About this item
:
:18.02 g/mol
Storage Temperature:Omgivande
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332