Desiccant replacement beads, Secador®, SP Bel-Art
Catalog # BELAF420740020
Supplier: Bel-Art Products, a Part of SP
Specifications
- Beskrivning:Replacement silica gel beads for Secador® auto-desiccator modules
- Förp.:25 g
Specifications
About this item
These colour-indicating silica gel beads are for use in refilling the Secador® auto-desiccator module which automatically regenerates them for thousands of cycles.
- Silica gel absorbs moisture from the air and is inert, non toxic and non corrosive
- Beads turn from blue to light pink indicating that the silica gel is saturated and cannot absorb additional moisture
- Beads turn pink/dark brown and fail to regain their blue colour when it is time to replace the beads with a fresh supply