Teststrips, Peroxidas, MQuant®, Supelco®
Katalog # 1.17828.0001
Leverantör: Merck
CAS-nummer:
Specifikationer
- Kemisk metod:Enzymatic reaction
- Förp.:100 Tests
- Beskrivning:Peroxidas
- Typ:MQ
Specifikationer
Om denna produkt
MQuant® (MQ) test strips can be considered as a mobile laboratory on a few square millimeters of plastic strip. As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, thereby leading to potentially substantial savings in time and analytical resources. The strip is a biologically-degradable polyester foil and the low reagent content, in the reaction zones, assists problem-free waste disposal.
- Enkel hantering: doppa, vänta och jämför visuellt med färgskalan
- Säkert: inga extra reagenser krävs
- Resultat inom 2 minuter
- Kan användas både på laboratoriet och ute i fält