Test kits, acidity, model AC-DT
Catalog # HACH2064000
Supplier: Hach
CAS Number:
Specifications
- Kemisk metod:Digital Titrator/Sodium Hydroxide
- Förp.:1 KIT
- Beskrivning:Acidity test
Specifications
About this item
This kit is used to determine acidity concentrations.
- Digital titrator offers higher levels of precision and accuracy than drop count procedures
- Rugged titration device is ideal for field use, with no glass burettes required
Acidity is usually caused by the presence of mineral acids, salts of strong acids or free carbon dioxide. In surface or ground waters, acidity may form industrial pollution which in excessive levels cause corrosion and can be detrimental to fish.
Please note some of the reagents supplied in the kits are hazardous and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Delivery: Includes digital titrator, reagents and labware, manual and carrying case.