Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Elektrokemimätare
pH-mätare
Accessories for Lab 745/845/945 meters
Accessories for Lab 745/845/945 meters
Accessories for Lab 745/845/945 meters
Leverantör:
SI ANALYTICS
Certifikat
Expandera bilden
Accessories for Lab 745/845/945 meters
Leverantör:
SI ANALYTICS
Om denna produkt