VWR® CHEMI only, Chemiluminescence Imaging System
Catalog # 730-1471
Supplier: VWR Collection
Specifications
- Beskrivning:VWR® CHEMI only
- W×D×H:370×470×460 mm
- Lins:Fixed f0,95
- Förp.:1
Specifications
About this item
The VWR® CHEMI only system is dedicated to chemiluminescence imaging. The system is built for high performance automation and features a next generation, high quantum efficiency CCD camera for even greater sensitivity. A very simple set-up process means a single click will automatically capture a quality image of any Western blot.
- Configured for maximum sensitivity to ensure even the faintest band on a blot can be captured
- High quantum efficiency camera (73% @ 425 nm) is very sensitive to low level light emissions
- Small footprint - takes up minimal bench space
Delivery: Each system is supplied complete with 4 MP 12-/16-bit CCD camera, VWR® Image Capture Software and two copies VWR® Gel Documentation Analysis Software.