Orders LinkContactUsLinkComponent
Dropping bottle, LDPE
Dropping bottle, LDPE
Supplier:   Hach
Dropping bottle, LDPE
Supplier:  Hach
Certificates
Dropping bottle, LDPE
Dropping bottle, LDPE
3 Options Available Below
Product Details & Documents
LDPE, translucent.
This dropping bottle is supplied without dropper plug and cap. Plugs and caps must be ordered separately.
Product Family Options
Product Information
  • Beskrivning
    Kapacitet
    Availability
    Price
  • Dropping bottle, without plug and cap
    59 ml
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items
  • Plug, 20 mm
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 99,00 kr
    99,00 kr
    1 * 6 Items
       99,00 kr
  • Cap, white, 20 mm
    Log in for availability
    Log in for availability
    1 * 6 Items
    1 * 6 Items - 80,00 kr
    80,00 kr
    1 * 6 Items
       80,00 kr