Media in bottles, wort agar
Catalog # DOEH840360782
Supplier: DOEHLER GROUP
CAS Number:
Specifications
- Förpackningstyp:Bottle
- Förp.:9
- Beskrivning:Wort agar, glas bottle
- Storlek:250 ml
- Fyllning:250 ml
Specifications
About this item
Agar for isolation yeast and moulds.
- For membrane filtration, pour plate and streaking method
- High selectivity