Immobiline™ DryPlate for IEF
Katalog # 80-1128-28
Leverantör: Cytiva
Specifikationer
- Beskrivning:Immobiline™ DryPlate pH 4,0 – 7,0
- Förp.:3
Specifikationer
Om denna produkt
Obtain reproducible IEF separations with dry, precast gels containing immobilized pH gradients.
- Choose from narrow (1 pH unit) and medium (3 pH units) pH ranges
- Handle stable, plastic-backed gels with confidence
Immobiline™ DryPlate precast gels contain preformed, immobilized, non-drifting pH gradients. The gels are rehydrated to a thickness of 0.5 mm in the reswelling cassette accessory before use.
Immobiline™ technology produces a stable pH gradient, allowing high voltages and resolution over narrow pH ranges, even for low mobility proteins that require long focusing times.
Ordering information: Immobiline™ sample application strips include 52 samples, 5–20µL and instructions.
Delivery: Kit includes 50× filter paper, 3× experimental result form instructions.