Hand crimper
Catalog # PERCN9302785
Supplier: PerkinElmer
Specifications
- Beskrivning:Hand crimper for chromatography vials, 20 mm
- Förp.:1
Specifications
About this item
Hand crimper for GC Headspace vials or LC large volume vials. It ensures sample integrity, particularly those containing volatile compounds.